Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249CE8A

J & W Pharmlab LLC

Isoquinonline-8-boronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
255.12
MDL number:

Iupac Name

8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline

Inchi Code

1S/C15H18BNO2/c1-14(2)15(3,4)19-16(18-14)13-7-5-6-11-8-9-17-10-12(11)13/h5-10H,1-4H3

InChI key

MSYQUACKTLEYFY-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

light yellow solid

Product Link

https://www.jwpharmlab.com/home/product/32R0045P

MSDS

https://www.jwpharmlab.com/home/product/32R0045P


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service