Skip to Content

We are planning system maintenance on January 25, 2025, from 1:00 PM to 4:00 PM CST. This will impact both web and offline transactions, including online orders, quotes, price and availability checks, and order status inquiries. We apologize for any inconvenience.

Merck
All Photos(1)

Key Documents

ENAH97BFAB9E

Enamine

1-(2-ethenylpyrrolidin-1-yl)ethan-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€564.81
1 G
€1,146.30
2.5 G
€2,226.21

Vendor SKU: BB20-7782A-250MG

€564.81


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
€564.81
1 G
€1,146.30
2.5 G
€2,226.21

About This Item

CAS Number:
Molecular Weight:
139.2
MDL number:

Vendor SKU: BB20-7782A-250MG

€564.81


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-(2-vinylpyrrolidin-1-yl)ethan-1-one

Inchi Code

1S/C8H13NO/c1-3-8-5-4-6-9(8)7(2)10/h3,8H,1,4-6H2,2H3

InChI key

FZUYNYZHQBWLBA-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-7110635

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service