Skip to Content
Merck
All Photos(1)

Key Documents

ENAH961CE34B

Enamine

3-butanoylbenzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€418.10
250 MG
€578.00
500 MG
€886.10
1 G
€1,122.70

Vendor SKU: EN300-8237810-100MG

€418.10


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
€418.10
250 MG
€578.00
500 MG
€886.10
1 G
€1,122.70

About This Item

CAS Number:
Molecular Weight:
173.21
MDL number:

Vendor SKU: EN300-8237810-100MG

€418.10


Ships in 14 days - from Aldrich Partner.

mp

65-66

Iupac Name

3-butyrylbenzonitrile

Inchi Code

1S/C11H11NO/c1-2-4-11(13)10-6-3-5-9(7-10)8-12/h3,5-7H,2,4H2,1H3

InChI key

OMDHPOSJTIUROC-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-8237810

Melting Point

65-66

MSDS

https://www.enaminestore.com/catalog/EN300-8237810


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service