ENAH961CDEB3
Enamine
5-tert-butyl 2-ethyl 3-bromo-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-2,5-dicarboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
5-tert-butyl 2-ethyl 3-bromo-6,7-dihydropyrazolo[1,5-a]pyrazine-2,5(4H)-dicarboxylate
Inchi Code
1S/C14H20BrN3O4/c1-5-21-12(19)11-10(15)9-8-17(6-7-18(9)16-11)13(20)22-14(2,3)4/h5-8H2,1-4H3
InChI key
DAAXIXUFHZXYGF-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-24888357
MSDS
https://www.enaminestore.com/catalog/EN300-24888357
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service