Skip to Content
Merck
All Photos(1)

Key Documents

ENAH38A2BC29

Enamine

(2-methyloxiran-2-yl)methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
88.11
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

[(2S)-2-methyl-2-oxiranyl]methanol

Inchi Code

1S/C4H8O2/c1-4(2-5)3-6-4/h5H,2-3H2,1H3

InChI key

AAVHHYWBSVSPPN-UHFFFAOYSA-N

storage temp.

-10

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-214874

Flash Point

62

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service