Skip to Content
Merck

ENAH3045D1B5

Enamine

N-[(4-bromothiophen-2-yl)methyl]-2-fluoroaniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
286.17
MDL number:

Pricing and availability is not currently available.

Iupac Name

N-[(4-bromo-2-thienyl)methyl]-2-fluoroaniline

Inchi Code

1S/C11H9BrFNS/c12-8-5-9(15-7-8)6-14-11-4-2-1-3-10(11)13/h1-5,7,14H,6H2

InChI key

BVBCHPOSHPXSJO-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-165654

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service