ENA429361122
Enamine
2-(cyclopentylamino)-1,3-thiazole-4-carboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-(cyclopentylamino)-1,3-thiazole-4-carboxylic acid
Inchi Code
1S/C9H12N2O2S/c12-8(13)7-5-14-9(11-7)10-6-3-1-2-4-6/h5-6H,1-4H2,(H,10,11)(H,12,13)
InChI key
SHPOJMUZHLUQPG-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-191866
MSDS
https://www.enaminestore.com/catalog/EN300-191866
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service