ENA024295534
Enamine
4-[4-(difluoromethoxy)phenyl]-5-[(piperidin-1-yl)methyl]-4H-1,2,4-triazole-3-thiol
Select a Size
Vendor SKU: EN300-09498-100MG
€165.90
Select a Size
About This Item
Vendor SKU: EN300-09498-100MG
€165.90
Iupac Name
4-[4-(difluoromethoxy)phenyl]-5-(1-piperidinylmethyl)-4H-1,2,4-triazol-3-yl hydrosulfide
Inchi Code
1S/C15H18F2N4OS/c16-14(17)22-12-6-4-11(5-7-12)21-13(18-19-15(21)23)10-20-8-2-1-3-9-20/h4-7,14H,1-3,8-10H2,(H,19,23)
InChI key
HAAHJJOVKVCGPY-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-09498
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service