Skip to Content
Merck
All Photos(1)

Key Documents

CO1H324A8FEC

CombiPhos Catalysts Inc

1-(5-AMINOPYRIDIN-2-YL)PIPERIDIN-2-OL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
193.25

Iupac Name

1-(5-aminopyridin-2-yl)piperidin-2-ol

Inchi Code

1S/C10H15N3O/c11-8-4-5-9(12-7-8)13-6-2-1-3-10(13)14/h4-5,7,10,14H,1-3,6,11H2

InChI key

CPVZFLPJCVLRBA-UHFFFAOYSA-N

Purity

95%

Country of Origin

US

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service