Skip to Content
Merck
All Photos(1)

Key Documents

COM456878695

Combi-Blocks, Inc.

trans-(4-Propylcyclohexyl)cyclohex-1-enylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
332.33
MDL number:

Pricing and availability is not currently available.

Iupac Name

(S)-4,4,5,5-tetramethyl-2-(4'-propyl-[1,1'-bi(cyclohexan)]-3-en-4-yl)-1,3,2-dioxaborolane

Inchi Code

1S/C21H37BO2/c1-6-7-16-8-10-17(11-9-16)18-12-14-19(15-13-18)22-23-20(2,3)21(4,5)24-22/h14,16-18H,6-13,15H2,1-5H3/t16?,17?,18-/m1/s1

InChI key

QSIBNMWSVJTKLV-DAWZGUTISA-N

storage temp.

Freezer

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?PN-6443

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?PN-6443


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service