Skip to Content
Merck

COM448614801

Combi-Blocks, Inc.

4-[(3-Fluorophenyl)(hydroxy)methyl]phenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
246.05
MDL number:

Pricing and availability is not currently available.

Iupac Name

4-[(3-fluorophenyl)(hydroxy)methyl]phenylboronic acid

Inchi Code

1S/C13H12BFO3/c15-12-3-1-2-10(8-12)13(16)9-4-6-11(7-5-9)14(17)18/h1-8,13,16-18H

InChI key

KQKCINWLQAXWDS-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA062066HPA037601HPA026859
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

Gene Information

human ... INPP1(3628)

Gene Information

human ... ENPP1(5167)

Gene Information

human ... INPPL1(3636)

Gene Information

human ... MINPP1(9562)

storage temp.

Refrigerated

Purity

97%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-4466

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-4466


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service