CIAH999A8198
ChemScene LLC (Preferred Partner)
trans-2-(Ethoxycarbonyl)cyclopropanecarboxylic acid
About This Item
bp
266.8+-33.0 C at 760 mmHg
Iupac Name
(1R,2R)-2-(ethoxycarbonyl)cyclopropanecarboxylic acid
Inchi Code
1S/C7H10O4/c1-2-11-7(10)5-3-4(5)6(8)9/h4-5H,2-3H2,1H3,(H,8,9)/t4-,5-/m1/s1
InChI key
BRVQFDJETHFEQY-RFZPGFLSSA-N
storage temp.
Purity
Country of Origin
Physical Form
Product Link
Boiling point
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0100043.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service