CH6371387898
Chem Impex International, Inc.
N-(1,2-Dimethylpropyl)cyclopropanamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
N-(1,2-dimethylpropyl)cyclopropanamine
Inchi Code
1S/C8H17N/c1-6(2)7(3)9-8-4-5-8/h6-9H,4-5H2,1-3H3
InChI key
NMZNLAPAPRESJL-UHFFFAOYSA-N
storage temp.
Store at 0-8C
Purity
95%
Shipping Temperature
RT
Physical Form
Liquid
Product Link
https://www.chemimpex.com/product/productinfo/n-1-2-dimethylpropyl-cyclopropanamine/16798?cid=0
MSDS
https://www.chemimpex.com/product/productinfo/n-1-2-dimethylpropyl-cyclopropanamine/16798?cid=0
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service