Skip to Content
Merck
All Photos(1)

Key Documents

860563C

Avanti

18:1-12:0 Biotin PC

Avanti Research - A Croda Brand 860563C

Synonym(s):

1-oleoyl-2-[12-biotinyl(aminododecanoyl)]-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€629.00

€629.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€629.00

About This Item

Empirical Formula (Hill Notation):
C48H89N4O10PS
CAS Number:
Molecular Weight:
945.28
UNSPSC Code:
12352211
NACRES:
NA.25

€629.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 1 mL (860563C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860563C

concentration

1 mg/mL (860563C-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)C)=O)(OC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI key

ANVUOOKUJFVAPJ-KKWOEEAUSA-N

Application

18:1-12:0 Biotin PC may be used in lipid blot analysis to study Niemann-Pick C2 (NPC2)-lipid interactions.[1] It may also be used in the preparation of liposomes.[2]

Packaging

5 mL Clear Glass Sealed Ampule (860563C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular cholesterol trafficking is dependent upon NPC2 interaction with lysobisphosphatidic acid
McCauliff LA, et al.
eLife, 8 (2019)
The Rab11-binding protein RELCH/KIAA1468 controls intracellular cholesterol distribution
Sobajima T, et al.
The Journal of Cell Biology, 217(5), 1777-1796 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service