Skip to Content
Merck
All Photos(1)

Key Documents

857411P

Avanti

IG4

Avanti Research - A Croda Brand, powder

Synonym(s):

4-fluoro-N-((2S,3R)-3-hydroxy-1-oxo-1-(tridecylamino)butan-2-yl)-1H-indole-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40FN3O3
CAS Number:
Molecular Weight:
461.61
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857411P-1mg)
pkg of 1 × 5 mg (857411P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@H](O)[C@H](NC(C(N1)=CC2=C1C=CC=C2F)=O)C(NCCCCCCCCCCCCC)=O

General description

IG4 is an analog of ceramide.

Packaging

5ML Glass Amber Screw Cap Vial (857411P-1MG)
5ML Glass Amber Screw Cap Vial (857411P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide mediates FasL-induced caspase 8 activation in colon carcinoma cells to enhance FasL-induced cytotoxicity by tumor-specific cytotoxic T lymphocytes
Coe G L, et al.
Scientific reports, 6(1), 1-15 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service