Skip to Content
Merck
All Photos(2)

Documents

810142P

Avanti

4ME 16:0 NBD PE (NBD-DPhPE)

1,2-diphytanoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt), powder

Synonym(s):

NBD-DPhPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H94N5O11P
CAS Number:
Molecular Weight:
984.29
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810142P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810142P

shipped in

dry ice

storage temp.

−20°C

General description

NBD-DPhPE preferentially partitions, although not entirely, into the fluid phase of the cell membrane rather than the more ordered cholesterol and sphingolipid-enriched microdomain known as a lipid "raft."NBD-DSPE preferentially partitions, although not entirely, into the more ordered cholesterol and sphingolipid-enriched microdomain known as a lipid "raft." - Personal communication, Dr. Erwin London, SUNY-Stony Brook

Packaging

5 mL Amber Glass Screw Cap Vial (810142P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Quinn
Progress in lipid research, 49(4), 390-406 (2010-05-19)
Domains in cell membranes are created by lipid-lipid interactions and are referred to as membrane rafts. Reliable isolation methods have been developed which have shown that rafts from the same membranes have different proteins and can be sub-fractionated by immunoaffinity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service