Skip to Content
Merck
All Photos(1)

Key Documents

ATEH75DC0178

AstaTech, Inc.

3-ETHOXYPROP-1-YNE

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
84.12
MDL number:

Iupac Name

3-ethoxyprop-1-yne

Inchi Code

1S/C5H8O/c1-3-5-6-4-2/h1H,4-5H2,2H3

InChI key

ADJMUEKUQLFLQP-UHFFFAOYSA-N

Purity

95%

Physical Form

Colorless to Yellow Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=172697

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C90311


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service