Skip to Content
Merck
All Photos(1)

Key Documents

ATEH0DDC42C9

AstaTech, Inc.

METHYL 5-(TERT-BUTOXYCARBONYL(METHYL)AMINO)THIOPHENE-2-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
271.34
MDL number:

Iupac Name

methyl 5-((tert-butoxycarbonyl)(methyl)amino)thiophene-2-carboxylate

Inchi Code

1S/C12H17NO4S/c1-12(2,3)17-11(15)13(4)9-7-6-8(18-9)10(14)16-5/h6-7H,1-5H3

InChI key

BZKQHHLHGDCHQW-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=121998

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=12966


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service