ATE514318914
AstaTech, Inc.
TERT-BUTYL 9-CYANO-3-AZASPIRO[5.5]UNDECANE-3-CARBOXYLATE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
tert-butyl 9-cyano-3-azaspiro[5.5]undecane-3-carboxylate
Inchi Code
1S/C16H26N2O2/c1-15(2,3)20-14(19)18-10-8-16(9-11-18)6-4-13(12-17)5-7-16/h13H,4-11H2,1-3H3
InChI key
MZPKZVLAVUVOPO-UHFFFAOYSA-N
storage temp.
Room temperature
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
Pale-yellow to Yellow-brown Liquid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=83460
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=F14353
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service