Skip to Content
Merck
All Photos(1)

Key Documents

ATE448436930

AstaTech, Inc.

METHYL 4-AMINO-2-CHLORO-3-IODOBENZOATE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€724.29
1 G
€1,388.59

Vendor SKU: SC5027-250MG

€724.29


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
€724.29
1 G
€1,388.59

About This Item

CAS Number:
Molecular Weight:
311.51
MDL number:

Vendor SKU: SC5027-250MG

€724.29


Ships in 14 days - from Aldrich Partner.

Iupac Name

methyl 4-amino-2-chloro-3-iodobenzoate

Inchi Code

1S/C8H7ClINO2/c1-13-8(12)4-2-3-5(11)7(10)6(4)9/h2-3H,11H2,1H3

InChI key

UPSLUSZCJZGFEL-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

97%

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=61112

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=SC5027


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service