Skip to Content
Merck
All Photos(1)

Key Documents

ATE067529686

AstaTech, Inc.

N-(4-BROMOPHENYL-N-(3-CYANOPHENYL)AMINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
273.13
MDL number:

Iupac Name

3-(4-bromoanilino)benzonitrile

Inchi Code

1S/C13H9BrN2/c14-11-4-6-12(7-5-11)16-13-3-1-2-10(8-13)9-15/h1-8,16H

InChI key

KNBAHWVBIOSXQS-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

97%

Shipping Temperature

Normal

Product Link

http://astatechinc.com/CPSResult.php?CRNO=5297

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=56995


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service