Skip to Content
Merck
All Photos(1)

Key Documents

APOH316CDD2E

Apollo Scientific Ltd

Di-(tert-butyl) diazene-1,2-dicarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€59.80
100 G
€88.40

Vendor SKU: OR5753-25G

€59.80


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
25 G
€59.80
100 G
€88.40

About This Item

CAS Number:
Molecular Weight:
230.26
MDL number:

Vendor SKU: OR5753-25G

€59.80


Ships in 14 days - from Aldrich Partner.

bp

89-92

Iupac Name

di(tert-butyl) (E)-1,2-diazenedicarboxylate

Inchi Code

1S/C10H18N2O4/c1-9(2,3)15-7(13)11-12-8(14)16-10(4,5)6/h1-6H3

InChI key

QKSQWQOAUQFORH-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/di-tert-butyl-diazene-12-dicarboxylate

Boiling point

89-92

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/di-tert-butyl-diazene-12-dicarboxylate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service