Skip to Content
Merck
All Photos(1)

Key Documents

APOH03CD9B00

Apollo Scientific Ltd

2,4-Dibromo-5-methoxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
293.94
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

2,4-dibromo-5-methoxybenzaldehyde

Inchi Code

1S/C8H6Br2O2/c1-12-8-2-5(4-11)6(9)3-7(8)10/h2-4H,1H3

InChI key

YNFYLBAXTVUPKS-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Product Link

https://store.apolloscientific.co.uk/product/24-dibromo-5-methoxybenzaldehyde

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/24-dibromo-5-methoxybenzaldehyde


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service