APO455835537
Apollo Scientific Ltd
3,3'-Bis(trifluoromethyl)benzophenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
98-102
Iupac Name
bis[3-(trifluoromethyl)phenyl]methanone
Inchi Code
1S/C15H8F6O/c16-14(17,18)11-5-1-3-9(7-11)13(22)10-4-2-6-12(8-10)15(19,20)21/h1-8H
InChI key
POOXOHISLGOAEF-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/33-bistrifluoromethylbenzophenone
Synonym
Bis[3-(trifluoromethyl)phenyl]methanone
Boiling point
98-102
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/33-bistrifluoromethylbenzophenone
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service