Skip to Content
Merck
All Photos(1)

Key Documents

APO455831988

Apollo Scientific Ltd

Methyl 3-aminophenylacetate hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€66.40
5 G
€124.70
25 G
€349.10

Vendor SKU: OR59469-1G

€66.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
€66.40
5 G
€124.70
25 G
€349.10

About This Item

CAS Number:
Molecular Weight:
201.65
MDL number:

Vendor SKU: OR59469-1G

€66.40


Ships in 14 days - from Aldrich Partner.

Iupac Name

methyl (3-aminophenyl)acetate hydrochloride

Inchi Code

1S/C9H11NO2.ClH/c1-12-9(11)6-7-3-2-4-8(10)5-7;/h2-5H,6,10H2,1H3;1H

InChI key

OOGVJPZOIXNNHQ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/methyl-3-aminophenylacetate-hydrochloride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/methyl-3-aminophenylacetate-hydrochloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service