Skip to Content
Merck
All Photos(1)

Key Documents

APO444780077

Apollo Scientific Ltd

4-tert-Butyl-1,3-thiazole-2-carboxaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
169.25
MDL number:

Iupac Name

4-tert-butyl-1,3-thiazole-2-carbaldehyde

Inchi Code

1S/C8H11NOS/c1-8(2,3)6-5-11-7(4-10)9-6/h4-5H,1-3H3

InChI key

ZSICFKHOZSNTLK-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/4-tert-butyl-13-thiazole-2-carboxaldehyde

Flash Point

50-52/2mm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-tert-butyl-13-thiazole-2-carboxaldehyde


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service