Key Documents
Z687782
Replacement NMR needle with cone tip for eVol® electronic syringe
for 50 μL, L × O.D. 18.0 cm × 0.63 mm, gauge size 23
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
gauge size
23
manufacturer/tradename
SGE 038038
L × O.D.
18.0 cm × 0.63 mm
General description
The eVol® NMR electronic syringe facilitates in-tube sample mixing and enables recovery of precious samples allowing them to be stored in alternative vessels (such as inexpensive vials) rather than being left in valuable tubes. The cone tip replacement needles come in multiple length and gauge options to fit your specific needs.
Legal Information
eVol is a registered trademark of SGE Analytical Science Pty Ltd.