WHA3668915
Whatman® cellulose chromatography papers
SG81 Sheets, W × L 460 mm × 570 mm
Synonym(s):
Whatman paper, Z742562, blotting papers, cellulose membranes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
cellulose membrane
sheets
grade
for chromatography
sterility
non-sterile
feature
CHR
packaging
pkg of 25 ea
manufacturer/tradename
Whatman 3668-915
Whatman Article No. 28419437 (US reference)
parameter
110 mm/30 min speed
W × L
460 mm × 570 mm
Looking for similar products? Visit Product Comparison Guide
General description
SG81 ion exchange sheets is a 0.27 mm combining cellulose and large pore silica gel for separations in which both partition and adsorption are important. A novel paper (0.27 mm thick) combining cellulose and large pore silica gel. Not charged, but binds polar molecules from less polar solvent. Suitable for separations in which both partition and adsorption are important, including the separation of phospholipids, steroids, phenols and dyes. Flow rate is 110 mm/30 min.
Features and Benefits
- Simultaneous development of multiple samples on the same sheet under identical conditions.
- Sequential development of the same samples with different solvents and/or different concentrations of the same solvent.
- Suitable for two-dimensional chromatography (change in direction of the solvent front) with possible improved resolution.
- Used in enzyme assays to separate product from reactant by charge or polarity.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Applied and environmental microbiology, 75(18), 5972-5980 (2009-07-28)
A Streptococcus mitis genomic DNA fragment carrying the SMT1224 gene encoding a putative beta-galactosidase was identified, cloned, and expressed in Escherichia coli. This gene encodes a protein 2,411 amino acids long with a predicted molecular mass of 268 kDa. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service