Skip to Content
Merck
All Photos(1)

Documents

P46202

Sigma-Aldrich

1-Piperidineacetaldehyde diethyl acetal

Synonym(s):

1-(2,2-Diethoxyethyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23NO2
CAS Number:
Molecular Weight:
201.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.443 (lit.)

bp

219-221 °C (lit.)

density

0.915 g/mL at 25 °C (lit.)

SMILES string

CCOC(CN1CCCCC1)OCC

InChI

1S/C11H23NO2/c1-3-13-11(14-4-2)10-12-8-6-5-7-9-12/h11H,3-10H2,1-2H3

InChI key

VSHHRRNJQBETGT-UHFFFAOYSA-N

Application

Reactant for:
  • Ethylene trimerisation
  • Synthesis of aryl-substituted piperidines by superacid activation of piperidones
  • Studies involving elimination reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service