Skip to Content
Merck
All Photos(4)

Key Documents

MP83502010

Sigma-Aldrich

MakroPor, silicon membrane

thickness 350 μm, pore diameter 8 μm, pore size 12 μm (interpore distance), size 20 mm × 20 mm, pkg of 10 pieces

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si
UNSPSC Code:
12352103
NACRES:
NA.23

description

FZ single crystal Si
Pore geometry: square arrangement

packaging

pkg of 10 pieces

size

20 mm × 20 mm

thickness

350 μm

pore size

12 μm (interpore distance)
8 μm pore diameter

Looking for similar products? Visit Product Comparison Guide

Application

Macro porous silicon is useful for applications requiring mechanical and chemical stability with high order of the pores. Our macro porous silicon membranes are manufactured from single crystal n-doped silicon wafers and possess pore diameter between 1 to 8 μm. The standard deviation of pore diameter and interpore distance for these porous silicon membrane materials is less than 5 %. These macro porous silicon finds applications in filtration, as platforms for multi-functional flow-through sensors, as fuels cells electrodes or micro reactors etc. They can also be used as templates for the fabrication of micro scale composites, such as tubes or wires by electrochemical deposition or by using polymer melts. We suggest using vacuum tweezers on the front side or polymer tweezers on the side faces.

Legal Information

Product of SmartMembranes-GmbH

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service