Skip to Content
Merck
All Photos(1)

Key Documents

M50206

Sigma-Aldrich

Methyl 2-hydroxyisobutyrate

95%

Synonym(s):

Methyl α-hydroxyisobutyrate, Methyl 2-methyllactate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OH)COOCH3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1746607
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
form:
liquid
Assay:
95%

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.411 (lit.)

bp

137 °C (lit.)

density

1.023 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(C)(C)O

InChI

1S/C5H10O3/c1-5(2,7)4(6)8-3/h7H,1-3H3

InChI key

XYVQFUJDGOBPQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service