Skip to Content
Merck
All Photos(1)

Documents

GF51628226

High permeability magnetic alloy, Ni80Fe15Mo5

foil, 25mm disks, 0.012mm thickness

Synonym(s):

NI180220

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

foil (disc)

manufacturer/tradename

Goodfellow 516-282-26

diam. × thickness

25 mm × 0.012 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service