Skip to Content
Merck
All Photos(1)

Documents

CDS003337

Sigma-Aldrich

cupric carbonate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCuO3
CAS Number:
Molecular Weight:
123.55
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

reaction suitability

core: copper
reagent type: catalyst

SMILES string

[Cu++].[O-]C([O-])=O

InChI

1S/CH2O3.Cu/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

GEZOTWYUIKXWOA-UHFFFAOYSA-L

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmad Faizal Abdull Razis et al.
International journal of environmental research and public health, 17(23) (2020-12-06)
A total of 779 samples of edible nuts (melon seeds, watermelon seeds, pumpkin seeds, and cantaloupe seeds) from Southern Punjab (Pakistan), were collected during the summer and the winter seasons. The natural occurrence of aflatoxins (AFs) and vitamin E (tocopherols)
Frances M Wolber et al.
Nutrients, 8(2), 104-104 (2016-02-26)
Poultry feathers, consisting largely of keratin, are a low-value product of the poultry industry. The safety and digestibility of a dietary protein produced from keratin (KER) was compared to a cysteine-supplemented casein-based diet in a growing rat model for four
A Zając et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 24(1), 11-20 (2018-10-26)
The copper phytate IP6Cu, IP6Cu2 and IP6Cu3 complexes were synthesized changing the phytate to metal mole ratio. The obtained products have been characterized by means of chemical and spectroscopic studies. Spectroscopic ATR/IR, FT-Raman, UV-Vis, EPR and magnetic measurements were carried
Amy McMillan et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 111, 356-362 (2017-11-28)
Aflatoxin exposure is an important public health concern in sub-Saharan Africa as well as parts of Latin America and Asia. In addition to hepatocellular carcinoma, chronic aflatoxin exposure is believed to play a role in childhood growth impairment. The most
Yoo Kim et al.
The Journal of nutritional biochemistry, 38, 125-133 (2016-10-14)
Previously, it was reported that conjugated linoleic acid (CLA) with exercise training potentially improved endurance capacity via the peroxisome proliferator-activated receptor δ (PPARδ)-mediated mechanism in mice. This study determined the role of exercise and/or CLA in endurance capacity and PPARδ-associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service