BAB604002100
SP Bel-Art® H-B Double-Safe™ PFA Coated, Liquid-In-Glass Thermometer
temp 0-300 °F, immersion level, Total Immersion
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
graduation 2 °F
immersion level Total Immersion
manufacturer/tradename
Bel-Art B60400-2100
temp measuring range
0-300 °F
accuracy: ±2 °F (above 212°F)
accuracy: ±2 °F
L
12 in. (300 mm)
Looking for similar products? Visit Product Comparison Guide
General description
These environmentally safe thermometers are PFA safety coated to keep liquid and glass contained in the event of breakage.
Features and Benefits
These environmentally safe thermometers are PFA safety coated to keep liquid and glass contained in the event of breakage.
- Non-toxic, biodegradable, Enviro-Safe® liquid, against lead-free, yellow back glass provides easy readability and fewer reading errors
- Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries
- Manufactured and tested by our USA quality team
- Traceable to NIST
- Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
- Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Double-Safe is a trademark of H-B Instrument Co.
Enviro-Safe is a registered trademark of H-B Instrument Co.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service