Skip to Content
Merck
All Photos(1)

Key Documents

A88409

Sigma-Aldrich

4,4′-Dimethoxybenzoin

95%

Synonym(s):

2-Hydroxy-4′-methoxy-2-(4-methoxyphenyl)acetophenone, p-Anisoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(CH3O)C6H4CH(OH)COC6H4-4-(OCH3)
CAS Number:
Molecular Weight:
272.30
Beilstein:
486157
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

mp

108-111 °C (lit.)

SMILES string

COc1ccc(cc1)C(O)C(=O)c2ccc(OC)cc2

InChI

1S/C16H16O4/c1-19-13-7-3-11(4-8-13)15(17)16(18)12-5-9-14(20-2)10-6-12/h3-10,15,17H,1-2H3

InChI key

LRRQSCPPOIUNGX-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Jackson et al.
Journal of applied microbiology, 122(4), 940-952 (2017-01-17)
The aim of this work was to isolate novel lignin-degrading organisms. Several pure cultures of bacteria that degrade lignin were isolated from bacterial consortia developed from decaying biomass. Among the isolates, Rhizobium sp. strain YS-1r (closest relative of Rhizobium petrolearium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service