Skip to Content
Merck
All Photos(2)

Key Documents

900328

Sigma-Aldrich

Poly(p-dioxanone-co-L-lactide)

viscosity 1.5 dL/g 

Synonym(s):

PDO-PLLA, PDS-PLLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C4H6O3)m(C6H8O4)nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

chips or granules

Quality Level

viscosity

1.5 dL/g

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Main features of p-dioxanone, glycolide and lactide based copolymers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service