Skip to Content
Merck
All Photos(1)

Key Documents

692387

Sigma-Aldrich

(R)-H8-BINAP

≥94%

Synonym(s):

(R)-(+)-2,2′-Bis(diphenylphospino)-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-binaphthyl, [(1R)-5,5′,6,6′,7,7′,8,8′-octahydro-[1,1′-binaphthalene]-2,2′-diyl]bis[diphenylphosphine]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H40P2
CAS Number:
Molecular Weight:
630.74
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥94%

form

powder

optical activity

[α]20/D +74°, c = 0.5 in toluene

InChI

1S/C44H40P2/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38/h1-12,19-26,29-32H,13-18,27-28H2

InChI key

ANSOKCGDSQQISA-UHFFFAOYSA-N

General description

The product is a diphenylphosphinobenzoic acid (DPPBA) based ligand for the palladium-catalyzed asymmetric allylic alkylation with a high degree of enantioselectivity.

Application

Catalytic ligand used for:
  • Enantioselective synthesis of dihydrobenzofurans and dihydronaphthofurans via olefin isomerization/enantioselective intramolecular Alder-ene reaction of enynes catalyzed by Rh
  • Preparation of axially chiral biaryl compounds by gold-catalyzed stereoselective intramolecular hydroarylation
  • Preparation of chiral 3-alkyl-substituted indolines by tandem condensation-asymmetric hydrogenation of indoles with aldehydes, catalyzed by Bronsted acids and palladium BINAP complexes
  • Stereoselective preparation of hydrindanes and decalins containing up to four contiguous stereocenters via rhodium-catalyzed [2+2+2] cycloaddition of ene-allenes with allenes
  • Rhodium-catalyzed asymmetric formal olefination or cycloaddition of 1,3-dicarbonyl compounds with 1,6-diynes or 1,6-enynes
  • Stereoselective preparation of 2,3-substituted indolines via Pd-catalyzed hydrogenation of (α-hydroxyalkyl)indoles

Legal Information

Sold in collaboration with Takasago for research purposes only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 40(3) (2007)

Articles

We present an article concerning BINAP/SEGPHOS® Ligands and Complexes.

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service