Skip to Content
Merck
All Photos(1)

Documents

680524

Sigma-Aldrich

2-Chloro-5-(trifluoromethyl)phenylboronic acid

Synonym(s):

2-Chloro-5-(trifluoromethyl)benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BClF3O2
CAS Number:
Molecular Weight:
224.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

SMILES string

OB(O)c1cc(ccc1Cl)C(F)(F)F

InChI

1S/C7H5BClF3O2/c9-6-2-1-4(7(10,11)12)3-5(6)8(13)14/h1-3,13-14H

InChI key

YVMXEHZEYONARR-UHFFFAOYSA-N

Application

Reactant for:
  • Synthesis of aryl- and hetarylfurocoumarins via Suzuki reaction
  • Synthesis of Et canthinone-3-carboxylates from Et 4-bromo-6-methoxy-1,5-naphthyridine-3-carboxylate via a Pd-catalyzed Suzuki-Miyaura coupling and a Cu-catalyzed amidation reaction
  • Preparation of xanthines via one-pot coupling with diaminouracil
  • Pd-catalyzed Suzuki-Miyaura coupling with 8-bromo-1,5-naphthyridin-2-one
May contain varying amounts of anhydride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service