Skip to Content
Merck
All Photos(1)

Documents

670561

Sigma-Aldrich

(1R,2R)-(−)-Norpseudoephedrine

≥98.0% (NT)

Synonym(s):

(−)-Pseudonorephedrine, (1R,2R)-2-Amino-1-phenyl-1-propanol, (R,R)-α-(1-Aminoethyl)benzenemethanol, L-(−)-Norpseudoephedrine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥98.0% (NT)

form

solid

optical activity

[α]/D -34.0±2.0°, c = 3.5 in ethanol

storage temp.

2-8°C

SMILES string

C[C@@H](N)[C@H](O)c1ccccc1

InChI

1S/C9H13NO/c1-7(10)9(11)8-5-3-2-4-6-8/h2-7,9,11H,10H2,1H3/t7-,9+/m1/s1

InChI key

DLNKOYKMWOXYQA-APPZFPTMSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jitka Caslavska et al.
Electrophoresis, 37(5-6), 699-710 (2016-01-23)
Dynamic computer simulation data are compared for the first time with CE data obtained with a laboratory made system comprising an array of 8 contactless conductivity detectors (C(4) Ds). The experimental setup featured a 50 μm id linear polyacrylamide (LPA)
María José Ojeda-Montes et al.
Future medicinal chemistry, 9(18), 2129-2146 (2017-11-28)
Extracts from Ephedra species have been reported to be effective as antidiabetics. A previous in silico study predicted that ephedrine and five ephedrine derivatives could contribute to the described antidiabetic effect of Ephedra extracts by inhibiting dipeptidyl peptidase IV (DPP-IV).
Hyeon-Kyu Lee et al.
The Journal of organic chemistry, 77(12), 5454-5460 (2012-05-25)
Each of the enantiomers of both norephedrine and norpseudoephedrine were stereoselectively prepared from the common, prochiral cyclic sulfamidate imine of racemic 1-hydroxy-1-phenyl-propan-2-one by employing asymmetric transfer hydrogenation (ATH) catalyzed by the well-defined chiral Rh-complexes, (S,S)- or (R,R)-Cp*RhCl(TsDPEN), and HCO(2)H/Et(3)N as
John S Chappell et al.
Forensic science international, 195(1-3), 108-120 (2009-12-29)
A primary concern with the forensic analysis of the khat plant (Catha edulis) has been the need to preserve the principle psychoactive component, cathinone, which converts to the less-active substance, cathine, after harvesting. The loss of cathinone has serious legal
A Pokrywka et al.
International journal of sports medicine, 30(8), 569-572 (2009-04-22)
Pseudoephedrine (PSE) as a sympathomimetic is an ingredient of many proprietary medicines which are available on the medical market over the counter (OTC drugs). It can be converted to cathine (CATH, norpseudoephedrine) inside the body. Until the end of 2003

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service