mp
234-238 °C (lit.)
SMILES string
Clc1ccccc1-c2nc3ccccc3[nH]2
InChI
1S/C13H9ClN2/c14-10-6-2-1-5-9(10)13-15-11-7-3-4-8-12(11)16-13/h1-8H,(H,15,16)
InChI key
OILIYWFQRJOPAI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 196, 49-57 (2018-02-13)
Due to wide variety of biological and pharmacological activities of benzimidazole derivatives, the differences between 2-(2-Chlorophenyl)benzimidazole and 2-(4-Chlorophenyl) benzimidazole were researched by employing terahertz time-domain spectroscopy and density functional theory systematically. Although the only difference between their molecular configurations is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service