Skip to Content
Merck
All Photos(1)

Key Documents

473960

Sigma-Aldrich

(S)-3-Amino-1,2-propanediol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
91.11
Beilstein:
1719122
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −28±1°, c = 4 in 5 M HCl

impurities

<0.5 mol

refractive index

n20/D 1.483 (lit.)

bp

117-119 °C/0.4 mmHg (lit.)

density

1.175 g/mL at 25 °C (lit.)

SMILES string

NC[C@H](O)CO

InChI

1S/C3H9NO2/c4-1-3(6)2-5/h3,5-6H,1-2,4H2/t3-/m0/s1

InChI key

KQIGMPWTAHJUMN-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-3-Amino-1,2-propanediol-borate system can be used for the enantioseparation of monosaccharides, derivatized with either 1-phenyl-3-methyl-5-pyrazolone (PMP) or 8-aminonaphthalene-1,3,6-trisulfonate (ANT) by chiral ligand-exchange capillary electrophoresis (CE).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chiral resolution of monosaccharides as 1-phenyl-3-methyl-5-pyrazolone derivatives by ligand-exchange CE using borate anion as a central ion of the chiral selector.
Kodama S, et al.
Electrophoresis, 27(23), 4730-4734 (2006)
Simultaneous chiral resolution of monosaccharides as 8-aminonaphthalene-1, 3, 6-trisulfonate derivatives by ligand-exchange CE using borate as a central ion of the chiral selector.
Kodama S, et al.
Electrophoresis, 28(21), 3930-3933 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service