Skip to Content
Merck
All Photos(1)

Key Documents

469297

Sigma-Aldrich

N-Boc-L-phenylalaninal

97%

Synonym(s):

(S)-(−)-2-(tert-Butoxycarbonylamino)-3-phenylpropanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH[NHCO2C(CH3)3]CHO
CAS Number:
Molecular Weight:
249.31
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D −47°, c = 0.5 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

86-88 °C (lit.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

[H]C(=O)[C@H](Cc1ccccc1)NC(=O)OC(C)(C)C

InChI

1S/C14H19NO3/c1-14(2,3)18-13(17)15-12(10-16)9-11-7-5-4-6-8-11/h4-8,10,12H,9H2,1-3H3,(H,15,17)/t12-/m0/s1

InChI key

ZJTYRNPLVNMVPQ-LBPRGKRZSA-N

Gene Information

human ... CTSK(1513)

Application

Intermediate for the synthesis of hydroxyethylene dipeptide isosteres that have enzyme inhibition properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mordini, A. et al.
Tetrahedron Letters, 37, 5209-5209 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service