424811
2,4,6,8-Tetramethylcyclotetrasiloxane
95%
Synonym(s):
TMCTS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(HSiCH3O)4
CAS Number:
Molecular Weight:
240.51
Beilstein:
1775282
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.387 (lit.)
bp
134 °C (lit.)
mp
−69 °C (lit.)
density
0.986 g/mL at 25 °C (lit.)
SMILES string
C[SiH]1O[SiH](C)O[SiH](C)O[SiH](C)O1
InChI
1S/C4H16O4Si4/c1-9-5-10(2)7-12(4)8-11(3)6-9/h9-12H,1-4H3
InChI key
BQYPERTZJDZBIR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Compound undergoes hydrosilylation reactions.
Features and Benefits
Precursor for gate dielectrics in thin-film transistors (TFT), and is a component of photochemically formed SiOX monolayers on TiO.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
75.2 °F - closed cup
Flash Point(C)
24 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chemistry of Materials, 5, 1687-1687 (1993)
Dae-Yoon Kim et al.
Soft matter, 11(1), 58-68 (2014-11-20)
Unconventional star-shaped liquid crystals (abbreviated as SiLCs) were successfully synthesized by chemically connecting four cyanobiphenyl anisotropic mesogens to the periphery of a super-hydrophobic and ultra-flexible cyclic tetramethyltetrasiloxane ring with flexible hexyl chains. Based on the combined experimental techniques of differential
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service