411221
2,2′,6,6′-Tetrabromobisphenol A diallyl ether
99%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
99%
mp
118-120 °C (lit.)
SMILES string
CC(C)(c1cc(Br)c(OCC=C)c(Br)c1)c2cc(Br)c(OCC=C)c(Br)c2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Environmental science & technology, 54(10), 6262-6271 (2020-04-22)
Tetrabromobisphenol A (TBBPA), the most widely used brominated flame retardant, is reported to potentially possess risks in inducing obesity or obesity-related metabolic diseases. Considering the increasing environmental contamination of TBBPA analogues and their high structural similarities to the parent compound
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service