Skip to Content
Merck
All Photos(1)

Key Documents

406740

Sigma-Aldrich

Poly(propylene glycol) diglycidyl ether

average Mn ~640

Synonym(s):

Polyoxypropylene diglycidyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

mol wt

average Mn ~640

refractive index

n20/D 1.457

density

1.06 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Poly(propylene glycol) diglycidyl ether maybe used in the preparation of anti corrosive solvent free coatings. It has been reported to be used in the preparation of polymer gel and prepolymer polypropylene glycol diglycidyl ether diacrylate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a novel UV-curable prepolymer polypropyleneglycol diglycidyl ether diacrylate.
Huang B, et al.
Journal of Wuhan University of Technology-Materials Science Edition, 23(4), 495-498 (2008)
A study on the reactive diluent for the solvent-free epoxy anticorrosive coating
Li Y, et al.
Journal of Chemical and Pharmaceutical Research, 6(7), 2466-2469 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service