Skip to Content
Merck
All Photos(1)

Documents

402354

Sigma-Aldrich

2-Amino-6-chloro-3-formylchromone

97%

Synonym(s):

2-Amino-6-chloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6ClNO3
CAS Number:
Molecular Weight:
223.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

>300 °C (lit.)

SMILES string

[H]C(=O)C1=C(N)Oc2ccc(Cl)cc2C1=O

InChI

1S/C10H6ClNO3/c11-5-1-2-8-6(3-5)9(14)7(4-13)10(12)15-8/h1-4H,12H2

InChI key

GCFVPLUXLQEQJX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Wang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 76(3-4), 293-296 (2010-04-30)
A highly selective chemosensor based on fluoran dye for Fe(2+), 2'-anilino-3'-methyl-6'-dibuthylamino-N-((2'-(2''-ethylimino) methyl) naphthalen-2-ol) iso-indolin-1-one-fluoran (5), was designed and synthesized. The chemical structures of all the intermediates and the fluoran dye 5 are characterized by (1)H NMR, (13)C NMR, Ms and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service