Skip to Content
Merck
All Photos(1)

Key Documents

388157

Sigma-Aldrich

Titanium(IV) bis(ethyl acetoacetato)diisopropoxide

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3CH2OCOCH=C(O-)CH3]2Ti(OCH(CH3)2)2
CAS Number:
Molecular Weight:
424.31
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

technical grade

vapor pressure

0.33 psi ( 20 °C)

refractive index

n20/D 1.518 (lit.)

density

1.109 g/mL at 25 °C (lit.)

SMILES string

CCO\C(O[Ti](OC(C)C)(OC(C)C)O\C(OCC)=C/C(C)=O)=C\C(C)=O

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service