Skip to Content
Merck
All Photos(1)

Key Documents

356832

Sigma-Aldrich

Iron

wire, diam. 0.5 mm, ≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.9% trace metals basis

form

wire

reaction suitability

reagent type: catalyst
core: iron

resistivity

9.71 μΩ-cm

diam.

0.5 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Quantity

1 m (approximately 1.5 g)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomasz Pytrus et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 34(203), 263-268 (2013-07-31)
The aim of the study was to analyze the occurrence of anemia in children with newly diagnosed Crohn's disease and 3 months after the initiation of treatment depending on location and activity of the disease. 36 children aged from 2
Daily iron during pregnancy improves birth weight.
Jacqui Wise
BMJ (Clinical research ed.), 346, f3997-f3997 (2013-06-26)
Iron fortification and malaria risk in children.
Andrew M Prentice et al.
JAMA, 310(9), 914-915 (2013-09-05)
Elzbieta Krzesiek et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 34(203), 289-292 (2013-07-31)
Anemia is a frequent symptom of diseases of alimentary tract, also in children. Among others, inflammatory bowel disease, celiac disease and Helicobacter pylori are most often complicated by anemia. Not infrequently these disorders are accompanied by more than one type
Wanmohd Azhar Wbin-Wan-Ibrahim et al.
Pakistan journal of pharmaceutical sciences, 26(4), 823-826 (2013-07-03)
Heavy metals in cigarette tobacco such as iron may cause a serious damage on human health. Surveys showed that the accumulation of certain toxic heavy metals like cadmium, mercury, iron is very often due to the effect of smoking. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service