Skip to Content
Merck
All Photos(1)

Key Documents

323640

Sigma-Aldrich

Hexafluoropropylene oxide

98%

Synonym(s):

Hexafluoro-1,2-epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3F6O
CAS Number:
Molecular Weight:
166.02
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

bp

−42 °C (lit.)

SMILES string

FC(F)(F)C1(F)OC1(F)F

InChI

1S/C3F6O/c4-1(2(5,6)7)3(8,9)10-1

InChI key

PGFXOWRDDHCDTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended.

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Press. Gas Compr. Gas - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Schröder et al.
Scientific reports, 9(1), 2237-2237 (2019-02-21)
Bulk polytetrafluoroethylene (PTFE) possesses excellent chemical stability and dielectric properties. Indeed, thin films with these same characteristics would be ideal for electret applications. Previously, the electret properties of PTFE-like thin films produced by rf sputtering or plasma enhanced chemical vapor
Chuan-Hai Li et al.
Environmental science & technology, 53(6), 3287-3295 (2019-02-21)
Hexafluoropropylene oxide trimer acid (HFPO-TA) and hexafluoropropylene oxide dimer acid (HFPO-DA) have been used as perfluorooctanoic acid (PFOA) alternatives in the fluoropolymer industry for years. Their widespread environmental distribution, high bioaccumulation capability, and human exposure have caused great concern. Nevertheless

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service