Skip to Content
Merck
All Photos(1)

Key Documents

310565

Sigma-Aldrich

2-Phenylbenzoxazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO
CAS Number:
Molecular Weight:
195.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

102-104 °C (lit.)

SMILES string

c1ccc(cc1)-c2nc3ccccc3o2

InChI

1S/C13H9NO/c1-2-6-10(7-3-1)13-14-11-8-4-5-9-12(11)15-13/h1-9H

InChI key

FIISKTXZUZBTRC-UHFFFAOYSA-N

General description

2-Phenylbenzoxazole is present as a hydrophobic fluorospore in the fluorescent molecular sensors AS1-3. The synthesis and microbiological activity of a new series of 5-benzamido- and 5-phenylacetamidosubstituted-2-phenylbenzoxazole derivatives were studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiaobing Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(26), 7543-7552 (2007-06-22)
A series of amphiphilic intramolecular charge-transfer fluorescent molecular sensors AS1-3, equipped with a rod-shaped hydrophobic 2-phenylbenzoxazole fluorophore and a hydrophilic tetraamide Hg(2+)-ion receptor, have been prepared. These sensor molecules could be incorporated into the hydrophobic sodium dodecyl sulfate (SDS) micelle
E A Sener et al.
Farmaco (Societa chimica italiana : 1989), 55(5), 397-405 (2000-09-13)
The synthesis and microbiological activity of a new series of 5-benzamido- and 5-phenylacetamidosubstituted-2-phenylbenzoxazole derivatives (1-26) were described. The in vitro microbiological activity of the compounds was determined against Gram-positive, Gram-negative bacteria and the yeast Candida albicans in comparison with standard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service